missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD3 Antibody (AMPD3/901), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47691UV
This item is not returnable.
View return policy
Description
AMPD3 Monoclonal antibody specifically detects AMPD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AMPD3 | |
| Monoclonal | |
| DyLight 350 | |
| adenosine monophosphate deaminase (isoform E), adenosine monophosphate deaminase 3, AMP aminohydrolase, AMP deaminase 3, AMP deaminase isoform E, EC 3.5.4.6, Erythrocyte AMP deaminase, erythrocyte type AMP deaminase, erythrocyte-specific AMP deaminase, myoadenylate deaminase | |
| Recombinant full-length human AMPD3 protein (Uniprot: Q01432) | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 272 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| AMPD3/901 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction