missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiregulin Antibody (RM0151-7D17), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12036PE
This item is not returnable.
View return policy
Description
Amphiregulin Monoclonal antibody specifically detects Amphiregulin in Mouse samples. It is validated for Western Blot
Specifications
| Amphiregulin | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0151-7D17 | |
| Western Blot | |
| amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
| Mouse recombinant Amphiregulin | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Biology, Cytokine Research, Growth and Development, Signal Transduction | |
| 374 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction