Learn More
Amphotericin B, Streptomyces nodosus, Thermo Scientific™
Amphotericin B, CAS # 1397-89-3, is an antifungal that has been used against fungus caused by protozoan parasites of Leishmania genus.
Brand:  Thermo Scientific Alfa Aesar J61491.06
Product Code. 15464089
  • ONLINE EXCLUSIVE PRICE
    € 846.00
    € 828.98 / 5g
    Save € 17.02
    2% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Amphotericin B is an antibiotic that has antifungal activity and is produced mainly by Streptomyces nodosus
Applications
• It has antifungal activity against fungus caused by protozoan parasites of Leishmania genus by binding to an essential component of the fungal cell membrane and altering cell membrane permeability
• It can suppress bone marrow progenitor cells and induce Actinomycin D(CR001) sensitivity in drug resistant HELA cells in vitro
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS Documents