missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1/2 [p Thr172, p Thr183] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33579-20ul
This item is not returnable.
View return policy
Description
AMPK alpha 1/2 Monoclonal antibody specifically detects AMPK alpha 1/2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| AMPK alpha 1/2 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| AMPK, AMPKa1, PRKAA1 protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| A synthetic phosphorylated peptide around T183/T172 of human AMPK alpha 1/2 (NP_006242.5/NP_006243.2).,, Sequence:, LRTSC | |
| 20 μL | |
| Phospho Specific | |
| 5562 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction