missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1 Antibody (2B7), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22127APCCY7
This item is not returnable.
View return policy
Description
AMPK alpha 1 Monoclonal specifically detects AMPK alpha 1 in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry.
Specifications
| AMPK alpha 1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| PRKAA1 | |
| Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt No. Q13131] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| IgG1 |
| Flow Cytometry | |
| 2B7 | |
| Flow Cytometry | |
| AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5′-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5′-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| Mouse | |
| Ammonium sulfate precipitation | |
| Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control | |
| 5562.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction