missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-239G
This item is not returnable.
View return policy
Description
AMPK alpha 1 Polyclonal specifically detects AMPK alpha 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AMPK alpha 1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRKAA1 | |
| The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human 5'-AMP-activated Protein Kinase, catalytic Alpha-1 chain using the numbering given in entry BAA36547.1 | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control | |
| 5562 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction