missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMPK alpha-1 Recombinant Rabbit Monoclonal Antibody (23GB2685), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553542
This item is not returnable.
View return policy
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
Specifications
| AMPK alpha-1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Prkaa1 | |
| 5'-AMP-activated protein kinase alpha-1 catalytic subunit; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 63 kDa subunit; AAPK2; ACACA kinase; Acetyl-CoA carboxylase kinase; activated protein kinase; AI194361; AI450832; AL024255; AMP -activate kinase alpha 1 subunit; AMP activated protein kinase; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK; AMPK 63 kDa subunit; AMPK alpha 1; AMPK alpha-1 chain; AMPK subunit alpha-1; AMPK subunit pAMPK; AMPK1; AMPK2; AMPKa1; AMPKalpha1; AMPK-alpha-2 chain; C130083N04Rik; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; MGC33776; MGC57364; pAMPK; PRKAA; Prkaa1; PRKAA2; protein kinase; protein kinase AMP-activated catalytic subunit alpha 1; protein kinase, AMP-activated, alpha 1 catalytic subunit; tau-protein kinase PRKAA1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 105787, 5562, 65248 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2685 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54645, Q13131, Q5EG47 | |
| Prkaa1 | |
| A synthesized peptide derived from human AMPK alpha 1 (500-559AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction