missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 2 Antibody (5B4G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00
Specifications
| Antigen | AMPK alpha 2 |
|---|---|
| Clone | 5B4G1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
AMPK alpha 2 Monoclonal specifically detects AMPK alpha 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AMPK alpha 2 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| 5563 | |
| Purified recombinant fragment of human PRKAA2 (AA: 453-552) expressed in E. Coli. | |
| Primary | |
| 62.3 kDa |
| 5B4G1 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control | |
| PRKAA2 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title