Learn More
AMPK Alpha Mouse anti-Human, CoraLite 488, Clone: 1H8E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66536-100UL
Description
The AMPK Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AMPK Alpha. This antibody recognizes human, rat, mouse antigen. The AMPK Alpha antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| AMPK Alpha | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 5562 | |
| IgG1 |
| Immunofluorescence | |
| 1H8E1 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |