Learn More
AMPK Alpha Mouse anti-Human, CoraLite 488, Clone: 1H8E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66536-100UL
Product Code. 16830816
  • € 450.00 / 100µL
Description
The AMPK Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AMPK Alpha. This antibody recognizes human, rat, mouse antigen. The AMPK Alpha antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
AMPK Alpha
Monoclonal
CoraLite 488
100 μL
5562
IgG1
Immunofluorescence
1H8E1
Mouse
Primary
Human, Rat, Mouse