Learn More
AMPK alpha Mouse anti-Human, Mouse, Rat, Clone: 1H8E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66536-1-IG-20UL
This item is not returnable.
View return policy
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifications
| AMPK alpha | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P54645, Q13131, Q5EG47 | |
| PRKAA1 | |
| AMPK alpha Fusion Protein Ag24744 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1H8E1 | |
| Unconjugated | |
| PRKAA1 | |
| AMPK, AMPK alpha, AMPK subunit alpha 1, AMPK1, AMPKa1, PRKAA1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 105787, 5562, 65248 | |
| -20°C | |
| Liquid |