missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK beta 2/PRKAB2 Antibody (OTI4H4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45389
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
AMPK beta 2/PRKAB2 Monoclonal specifically detects AMPK beta 2/PRKAB2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| AMPK beta 2/PRKAB2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| 5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
| Mouse | |
| 30.1 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5565 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI4H4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| PRKAB2 | |
| Full length human recombinant protein of human PRKAB2 (NP_005390) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction