Learn More
Amprenavir, MedChemExpress
Product Code. 16493566
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493566 25 mg 25mg
16489827 5 mg 5mg
16453357 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16493566 Supplier Medchem Express Supplier No. HY17430
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Amprenavir (VX-478) is a HIV protease inhibitor (Ki=0.6 nM) used to treat HIV infection. Amprenavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.09 μM.
Amprenavir (VX-478) is a HIV protease inhibitor (Ki=0.6 nM) used to treat HIV infection. Amprenavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.09 μM.
Specifications
Chemical Name or Material Amprenavir
CAS 161814-49-9
Color White
Purity Grade Notes Research
Molecular Formula C25H35N3O6S
Quantity 25 mg
Synonym VX-478
Solubility Information DMSO : ≥ 100 mg/mL (197.77 mM)
SMILES O=C(O[C@@H]1COCC1)N[C@@H](CC2=CC=CC=C2)[C@H](O)CN(S(=O)(C3=CC=C(N)C=C3)=O)CC(C)C
Molecular Weight (g/mol) 505.63
Formula Weight 505.63
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only