Learn More
Amsacrine, MedChemExpress
Product Code. 16455748
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16455748
Brand: Medchem Express HY13551
MedChemExpress Amsacrine (m-AMSA; acridinyl anisidide) is an inhibitor of topoisomerase II, and acts as an antineoplastic agent which can intercalates into the DNA of tumor cells.
Amsacrine (m-AMSA
Specifications
Chemical Name or Material Amsacrine
CAS 51264-14-3
Color Red
Purity Grade Notes Research
Molecular Formula C21H19N3O3S
Quantity 10 mg
Synonym m-AMSA acridinyl anisidide
Solubility Information DMSO : 9.3 mg/mL (23.64 mM; Need ultrasonic and warming)
SMILES CS(=O)(NC1=CC=C(NC2=C(C=CC=C3)C3=NC4=CC=CC=C42)C(OC)=C1)=O
Molecular Weight (g/mol) 393.46
Formula Weight 393.46
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only