missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMSH/STAMBP Antibody (S07-6D1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19832-100ul
This item is not returnable.
View return policy
Description
AMSH/STAMBP Monoclonal antibody specifically detects AMSH/STAMBP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| AMSH/STAMBP | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S07-6D1 | |
| Unconjugated | |
| AMSHAssociated molecule with the SH3 domain of STAM, EC 3.1.2.15, EC 3.4.19.-, Endosome-associated ubiquitin isopeptidase, MGC126516, MGC126518, STAM binding protein, STAM-binding protein | |
| A synthetic peptide of human AMSH/STAMBP (Uniprot # O95630) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 10617 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction