missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576454
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The enzyme system for cleavage of glycine (glycine cleavage system; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). Glycine encephalopathy (GCE) may be due to a defect in any one of these enzymes.
Specifications
| AMT | |
| Polyclonal | |
| Unconjugated | |
| AMT | |
| aminomethyltransferase; aminomethyltransferase (glycine cleavage system protein T); aminomethyltransferase, mitochondrial; AMT; EG434437; GCE; GCST; GCVT; glycine cleavage system T protein; NKH | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 275, 306586, 434437 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48728, Q8CFA2 | |
| AMT | |
| Recombinant fusion protein of human AMT. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction