missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMTN Monoclonal Antibody (OTI1D6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527024
This item is not returnable.
View return policy
Description
AMTN Monoclonal Antibody for Western Blot, IHC (P)
Is a promoter of calcium phosphate mineralization, playing a critical role in the formation of the compact, mineralized, aprismatic enamel surface layer during the maturation stage of amelogenesis.
Specifications
| AMTN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UX39 | |
| AMTN | |
| Full length human recombinant protein of AMTN produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D6 | |
| Unconjugated | |
| AMTN | |
| amelotin; AMTN; RSTI689; UNQ689; UNQ689/PRO1329 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 401138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction