Learn More
Anabaseine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Anabaseine-2,4,5,6-d4 |
| CAS | 1020719-05-4 |
| Molecular Formula | C10 2H4 H8 N2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h3-4,6,8H,1-2,5,7H2/i3D,4D,6D,8D |
| SMILES | [2H]c1nc([2H])c(C2=NCCCC2)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(2,3,4,5-tetrahydropyridin-6-yl)pyridine |
| Molecular Weight (g/mol) | 164.24 |
| Formula Weight | 164.1252 |