missing translation for 'onlineSavingsMsg'
Learn More

(±) Anabasine, MedChemExpress

Product Code. 16415467
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16415467

Brand: Medchem Express HYW052144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (±) Anabasine is a biphasic muscle relaxant.

(±) Anabasine is a biphasic muscle relaxant.

Specifications

Chemical Name or Material (±) Anabasine
CAS 13078-04-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C10H14N2
Quantity 50 mg
Solubility Information DMSO : 130 mg/mL (801.33 mM; Need ultrasonic)
SMILES C1(C2NCCCC2)=CC=CN=C1
Molecular Weight (g/mol) 162.23
Formula Weight 162.23
Percent Purity 95.0%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.