Learn More
Ohaus™ Analog Microplate Vortex Mixer VXMPAL
Product Code. 15742239 Shop All Ohaus Products
Click to view available options
Orbit:
3.5 mm
Unit Size:
Each
Product Code. 15742239
Brand: Ohaus™ 30392155
Only null left
Add to basket
Add to basket
OHAUS™ Analog Microplate Vortex Mixer VXMPAL is built sturdy to provide a stable and reliable vortexing action.
  • The OHAUS Analogue Microplate Vortex Mixer is a variable speed analogue mixer designed for continuous operation
  • Designed for shaking microplates or tubes
  • High speed and small orbit is optimal for effectively mixing microplates
  • Choose from two modes of operation: 'Touch' mode which activates mixing when cup head is depressed, or 'On' mode when using the microplate attachments for continuous operation
  • Optional double microplate holder available
TRUSTED_SUSTAINABILITY
Specifications
Orbit 3.5 mm
Speed Range On mode: 300 to 2500 rpm, Touch mode: 300 to 3500 rpm
Electrical Requirements 230 V, 50/60 Hz, 0.13 A
Amperage 0.13 A
Holds Microplates
Product Type Microplate Vortex Mixer
Plug Type EU Plug
Dimensions (L x W) 26.70 x 13.70 cm
Height (English) 4.50 in.
Height (Metric) 11.4 cm
Length (English) 10.50 in.
Length (Metric) 26.70 cm
Width (English) 5.40 in.
Width (Metric) 13.70 cm
Description Vortex Mixer, Microplate, VXMPAL, EU
Model VXMPAL
No. of Positions 2