Learn More
ANAPC11 Mouse anti-Human, Clone: OTI1F2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506321
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of ′Lys-11′-linked polyubiquitin chains and, to a lower extent, the formation of ′Lys-48′- and ′Lys-63′-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex.Specifications
| ANAPC11 | |
| Monoclonal | |
| Unconjugated | |
| ANAPC11 | |
| APC11, Apc11p, HSPC214 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NYG5 | |
| ANAPC11 | |
| Full length human recombit protein of human ANAPC11 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.