missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANAPC2 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503407S
This item is not returnable.
View return policy
Description
ANAPC2 Monoclonal antibody specifically detects ANAPC2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ANAPC2 | |
| Monoclonal | |
| 0.38 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UJX6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29882 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| ANAPC2 | |
| 9230107K09Rik; AL024279; Anapc2; anaphase promoting complex subunit 2; anaphase-promoting complex subunit 2; Apc2; cyclosome subunit 2; Emi4; expressed during mesenchymal induction 4; Imi4; induced during mesenchymal induction 4; KIAA1406; mKIAA1406 | |
| Full length human recombinant protein of human ANAPC2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction