missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Anatabine-d3 (1,2,3,6-tetrahydropyridinly-3,3,4-d3), CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | rac-Anatabine-D3 (tetrahydropyridine-3,3,4-D3) |
| Molecular Formula | C10 D3 H9 N2 |
| Quantity | 0.01 g |
| Synonym | (±)-Anatabine-d3 (1,2,3,6-tetrahydropyridinly-3,3,4-d3), rac-Anatabine-D3 (tetrahydropyridine-3,3,4-D3), 1,2,3,6-Tetrahydro-2,3'-bipyridine-3,3,34-d3, (R,S)-Anatabine-3',3',4'-d3 |
| InChI Formula | InChI=1 S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-4,6,8,10,12 H,5,7H2/i1D,5D2 |
| SMILES | [2 H]C1=CCNC(c2cccnc2)C1([2 H])[2 H] |
| IUPAC Name | 3-(3,3,4-trideuterio-2,6-dihydro-1 H-pyridin-2-yl)pyridine |
| Molecular Weight (g/mol) | 163.2342 |
| Formula Weight | 163.1189 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction