Learn More
Abeba™ Anatom 2616 Shoes
Product Code. 15688177
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15688177 45 1 pair
15698167 36 1 pair
15608177 37 1 pair
15618177 38 1 pair
15628177 39 1 pair
15638177 40 1 pair
15648177 41 1 pair
15658177 42 1 pair
15668177 43 1 pair
15678177 44 1 pair
15698177 46 1 pair
15608187 47 1 pair
15618187 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15688177 Supplier Abeba™ Supplier No. 2616/45
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Microfiber, perforated
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Adjustable instep area
  • Double velcro fastening
  • Reflective material
  • Reflective material
  • Suitable for hygienically demanding areas
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color White
Material Microfiber
Size 45
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2616
Toe Material Steel