Learn More
Abeba™ Anatom 32131 Shoes
Product Code. 15679327
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
Unit Size:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15679327 37 1 pair
15669327 36 1 pair
15689327 38 1 pair
15699327 39 1 pair
15609337 40 1 pair
15619337 41 1 pair
15629337 42 1 pair
15639337 43 1 pair
15649337 44 1 pair
15659337 45 1 pair
15669337 46 1 pair
15679337 47 1 pair
15689337 48 1 pair
15835872 49 1 pair
15845872 50 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15679327 Supplier Abeba™ Supplier No. 32131/37
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color White
Material Microfiber
Size 37
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32131
Toe Material Steel