Learn More
Lenz Laborglasintrumente Anchor Stirrers with GL Thread Connection
Product Code. p-7222463
Change view
Click to view available options
Length (Metric):
540 mm
595 mm
640 mm
680 mm
700 mm
715 mm
For Use With (Equipment):
Lab Flange 100 LF
Lab Flange 150 LF
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) For Use With (Equipment) unitSize
11903911 700 mm Lab Flange 150 LF Each
11913911 715 mm Lab Flange 150 LF Each
11914011 640 mm Lab Flange 100 LF Each
11924011 680 mm Lab Flange 100 LF Each
11983901 540 mm Lab Flange 150 LF Each
11993901 595 mm Lab Flange 150 LF Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11903911 Supplier Lenz Laborglasintrumente Supplier No. 06321679
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Made of precision calibrated glass tube
  • Ground and polished bearing surface
  • For flange vessels
  • Made of DURAN™ tubing
Especificaciones
Diameter (Metric) 16 mm
For Use With (Equipment) Lab Flange 150 LF
Length (Metric) 700 mm
Thread Size 14 GL
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.