missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Andes Virus Nucleocapsid Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41254MFV450
This item is not returnable.
View return policy
Description
Andes Virus Nucleocapsid Polyclonal antibody specifically detects Andes Virus Nucleocapsid in Virus samples. It is validated for ELISA
Specifications
| Andes Virus Nucleocapsid | |
| Polyclonal | |
| ELISA | |
| ANDV, Nucleocapsid protein | |
| Antibody was raised against a 19 amino acid peptide near the amino terminus of the Andes virus nuclecapsid. The immunogen is located within amino acids 130 - 180 of Andes Virus Nucleocapsid. Amino Acid Squence: TRGRQTVKDNKGTRIRFKD | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 991232 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction