Learn More
Emma Safety Footwear Andes XD Safety Shoes
Features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
Brand: Emma Safety Footwear MM30556844
Additional Details : Weight : 1.69500kg
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Andes XD | |
Oiled Nubuck | |
44 |