missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen R/NR3C4 Antibody (DHTR/8698R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 311.00 - € 553.00
Specifications
| Antigen | Androgen R/NR3C4 |
|---|---|
| Clone | DHTR/8698R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684535
|
Novus Biologicals
NBP3-20564-20ug |
20 μg |
€ 311.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609854
|
Novus Biologicals
NBP3-20564-100ug |
100 μg |
€ 553.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Androgen R/NR3C4 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
| 1mM PBS with 0.05% BSA | |
| 367 | |
| IgG κ | |
| Protein A or G purified |
| DHTR/8698R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Synthetic peptide corresponding to residues (within aa 1-100) of human Androgen R/NR3C4. | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title