missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen R/NR3C4 Antibody (DHTR/9119R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23817-100ug
This item is not returnable.
View return policy
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Androgen R/NR3C4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Recombinant fragment (around aa1-200) of human Androgen R/NR3C4 protein (exact sequence is proprietary) | |
| 100 μg | |
| Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
| 367 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DHTR/9119R | |
| Unconjugated | |
| 1 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction