missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen R/NR3C4, p Ser210 Antibody (156C135.2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56603IR
This item is not returnable.
View return policy
Description
Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Androgen R/NR3C4 (p Ser213, p Ser210) | |
| Monoclonal | |
| DyLight 755 | |
| P10275 | |
| AR | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
| 367 | |
| Human, Mouse, Rat, Canine, Primate | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 156C135.2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In IGF-1 stimulated LNCaP cells (passage number 38), a ∼110 kDa band was observed. Please see Lin et al. 2003 for additional details. The serine phosphorylation site recognized by this antibody has been alternatively referred to Ser213 (Lee and Chang, 2003) and Ser210 (Lin et al, 2003). Variations in denotation can arise from how the sequence is counted in various GenBank accession numbers. The site is denoted as Ser213 in GenBank Accession No. A39248, which was used to design the immunogen. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction