missing translation for 'onlineSavingsMsg'
Learn More

Anethole trithione, MedChemExpress

Product Code. p-200101561
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16492256

Brand: Medchem Express HYB1223

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Anethole trithione, a sulfur heterocyclic choleretic, is a bile secretion-stimulating agent. Anethole trithione enhances salivary secretion and increases mAChRs, and can be used for dry mouth research.

Anethole trithione, a sulfur heterocyclic choleretic, is a bile secretion-stimulating agent. Anethole trithione enhances salivary secretion and increases mAChRs, and can be used for dry mouth research.

Specifications

Chemical Name or Material Anethole trithione
CAS 532-11-6
Color Orange
Purity Grade Notes Research
Molecular Formula C10H8OS3
Quantity 500 mg
Solubility Information DMSO : 12.5 mg/mL (52.01 mM; Need ultrasonic)
SMILES S=C1SSC(C2=CC=C(OC)C=C2)=C1
Molecular Weight (g/mol) 240.36
Formula Weight 240.36
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.