Learn More
Invitrogen™ ANGEL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140056
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EGDEPSSKRR KHQGVIKRNW EYICSHDKEK TKILGDKNVD PKCEDSENKF DFSVMSYNIL SQDLLEDNSH LYRHCRRPVL HWSFRFPNIL KEIK Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 84%.
ANGEL 2 is a member of the CCR4/nocturin family.
Specifications
| ANGEL2 | |
| Polyclonal | |
| Unconjugated | |
| ANGEL2 | |
| 2610307I21Rik; 5730410O10Rik; AI845692; angel homolog 2; angel homolog 2 (Drosophila); ANGEL2; Ccr4d; D1Ertd396e; D1Ertd654e; KIAA0759L; MNCb-4273; Protein angel homolog 2; RGD1305056 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90806 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VTE6 | |
| ANGEL2 | |
| Recombinant protein corresponding to Human ANGEL2. Recombinant protein control fragment (Product #RP-108797). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.