missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiogenin Antibody (MM0044-7M15), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-93512AF750
This item is not returnable.
View return policy
Description
Angiogenin Monoclonal specifically detects Angiogenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Angiogenin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ANG | |
| human recombinant Angiogenin protein | |
| 0.1 mL | |
| Primary | |
| This recognizes human Angiogenin in western blot. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0044-7M15 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Cancer | |
| 283.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction