missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiogenin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98773-100ul
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Angiogenin Polyclonal antibody specifically detects Angiogenin in Rat samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| Angiogenin | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat Angiogenin | |
| 100 μg | |
| Angiogenesis, Cancer | |
| 283 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion