missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (SN67-02), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-67664
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Rat samples. It is validated for Western Blot.
Specifications
| Angiopoietin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000 | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| Rabbit | |
| Protein A purified | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 284 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| SN67-02 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| ANGPT1 | |
| Synthetic peptide within Human Angiopoietin-1 aa 249-300/ 498. (SwissProt: Q15389 Human; SwissProt: O35460 Rat) | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction