missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-2 Antibody (MM0020-1F29), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-85467F
This item is not returnable.
View return policy
Description
Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human, Chicken samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Angiopoietin-2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ANGPT2 | |
| Human recombinant Angiopoietin-2 | |
| 0.1 mL | |
| Primary | |
| No cross reactivity with human Angiopoietin-1 | |
| Store at 4C in the dark. | |
| IgG2 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0020-1F29 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Cancer | |
| 285 | |
| Human, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction