Learn More
Angiopoietin-2 Antibody (Roche patent anti-ANG-2) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-27955JF525
Product Code. 30174435
  • € 431.00 / 0.10mL
Description
Angiopoietin-2 Monoclonal antibody specifically detects Angiopoietin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiopoietin-2
Monoclonal
Janelia Fluor 525
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand
ANGPT2
0.1 mL
Angiogenesis, Cancer
285
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Roche patent anti-ANG-2
50mM Sodium Borate
Human
Protein A purified