missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 1/ANGPTL1 Antibody (OTI1B2), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71414JF646
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 1/ANGPTL1 Monoclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Angiopoietin-like Protein 1/ANGPTL1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANGPTL1 | |
| Recombinant protein fragment corresponding to amino acids 24-491 of human Angiopoietin-like Protein 1/ANGPTL1 (NP_004664) produced in E.coli. | |
| 0.1 mL | |
| 9068.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1B2 | |
| Western Blot, Immunohistochemistry | |
| ANG-3, ANG3angioarrestin, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, AngY, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction