missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 3/ANGPTL3 Antibody (23D8), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-60244G
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Monoclonal antibody specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Angiopoietin-like Protein 3/ANGPTL3 | |
| Monoclonal | |
| DyLight 488 | |
| ANG-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, FHBL2 | |
| Recombinant human Angiopoietin-like Protein 3/ANGPTL3 (243-460aa) purified from E. coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 27329 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence | |
| 23D8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction