missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 6/ANGPTL6 Antibody (3H8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00083854-M07
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 6/ANGPTL6 Monoclonal antibody specifically detects Angiopoietin-like Protein 6/ANGPTL6 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Angiopoietin-like Protein 6/ANGPTL6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AGFAngiopoietin-related protein 5, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ARP5angiopoietin-related protein 6 | |
| ANGPTL6 (NP_114123, 211 a.a. ∽ 299 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GSTSDTSRMLDPAPEPQRDQTQRQQEPMASPMPAGHPAVPTKPVGPWQDCAEARQAGHEQSGVYELRVGRHVVSVWCEQQLEGGGWTVI | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3H8 | |
| Western Blot 1:500, ELISA | |
| NP_114123 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 83854 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction