missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like protein 8/Betatrophin Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41247IR
This item is not returnable.
View return policy
Description
Angiopoietin-like protein 8/Betatrophin Polyclonal antibody specifically detects Angiopoietin-like protein 8/Betatrophin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Angiopoietin-like protein 8/Betatrophin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599 | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of human Betatrophin. The immunogen is located within amino acids 40 - 90 of Betatrophin. Amino Acid Squence: VYRTTEGRLTKARNSL | |
| 0.1 mL | |
| Cell Biology | |
| 55908 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction