Learn More
Novus Biologicals™ Angiopoietin-like protein 8/Betatrophin Antibody (Regeneron patent anti-ANGPTL8) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28032-1mg
 View more versions of this product
Product Code. 30047412
  • € 920.00 / 1mg
Description
Angiopoietin-like protein 8/Betatrophin Monoclonal antibody specifically detects Angiopoietin-like protein 8/Betatrophin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiopoietin-like protein 8/Betatrophin
Monoclonal
Unconjugated
Q6UXH0
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
Regeneron patent anti-ANGPTL8
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599
ANGPTL8
1 mg
Cell Biology
55908