Learn More
Angiopoietin-like protein 8/Betatrophin Antibody (Regeneron patent anti-ANGPTL8), mFluor Violet 500 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28032MFV500
Product Code. 30114082
  • € 454.00 / 0.10mL
Description
Angiopoietin-like protein 8/Betatrophin Monoclonal antibody specifically detects Angiopoietin-like protein 8/Betatrophin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiopoietin-like protein 8/Betatrophin
Monoclonal
mFluor Violet 500 SE
C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599
ANGPTL8
0.1 mL
Cell Biology
55908
Store at 4°C in the dark.
IgG2
Flow Cytometry, ELISA, Functional Assay
Regeneron patent anti-ANGPTL8
50mM Sodium Borate
Human
Protein A purified