missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiostatin Antibody (MM0094-6F34), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12042AF532
This item is not returnable.
View return policy
Description
Angiostatin Monoclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.
Specifications
| Angiostatin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins | |
| 5340.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| MM0094-6F34 | |
| Western Blot | |
| PLG | |
| Human recombinant Angiostatin | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction