missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiostatin Antibody (MM0094-6F34), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12042JF646
This item is not returnable.
View return policy
Description
Angiostatin Monoclonal antibody specifically detects Angiostatin in Human samples. It is validated for Western Blot
Specifications
| Angiostatin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0094-6F34 | |
| Western Blot | |
| HAE4 | |
| Human recombinant Angiostatin | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 5340 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction