missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiotensin I Antibody (BGN/KA/4H), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62344G
This item is not returnable.
View return policy
Description
Angiotensin I Monoclonal specifically detects Angiotensin I in Human, Rat samples. It is validated for ELISA.
Specifications
| Angiotensin I | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AGT | |
| Synthetic peptide corresponding to amino acids 40-49 of the angiotensinogen precursor (Swiss-Prot entry P01019) conjugated to HMW carrier protein. | |
| 0.1 mL | |
| Primary | |
| NB100-62344 is specific for Angiotensin I and does not react with angiotensin II, III or angiotensinogen in ELISA. Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. Angiotensin I is important in the Renin-Angiotensin-Aldosterone System (RAAS). RAAS is responsible for the regulation of blood pressure and water retention in the body. Angiotensin I is produced from a reaction between the renal enzyme Renin and angiotensinogen. It is converted by Angiotensin Converting Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. | |
| Store at 4C in the dark. | |
| IgG2a |
| ELISA | |
| BGN/KA/4H | |
| ELISA | |
| alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 183 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction