missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiotensin II Antibody (KAA8), Alexa Fluor™ 647 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20123AF647
This item is not returnable.
View return policy
Description
Angiotensin II Monoclonal antibody specifically detects Angiotensin II in Human, Rat samples. It is validated for Western Blot
Specifications
| Angiotensin II | |
| Monoclonal | |
| Alexa Fluor 647 | |
| AGT, alpha-1 antiproteinase, antitrypsin, angiotensin I, ANHU, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8 | |
| Angiotensin II | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| KAA8 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 183 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction