missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Angiotensin II/III Antibody (Ang II E7 (BGN/KA/4L)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB100-62346
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Angiotensin II/III Monoclonal specifically detects Angiotensin II/III in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| Angiotensin II/III | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| AGT angiotensinogen (serpin peptidase inhibitor, clade A, member 8), angiotensin I, angiotensin II, ANHU, SERPINA8 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This product reacts with Angiotensin II and III. Negligible reactivity was observed with Angiotensin I and Angiotensinogen. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Ang II E7 (BGN/KA/4L) | |
| Unconjugated | |
| P01019 | |
| AGT | |
| Peptide sequence corresponding to angiotensin II, conjugated to a proprietary molecular weight carrier. Peptide sequence: DRVYIHPF | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 183 | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering