missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiotensin II/III Antibody (Ang II E7 (BGN/KA/4L)), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB100-62346G
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Angiotensin II/III Monoclonal specifically detects Angiotensin II/III in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| Angiotensin II/III | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AGT | |
| Peptide sequence corresponding to angiotensin II, conjugated to a proprietary molecular weight carrier. Peptide sequence: DRVYIHPF | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 183 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Ang II E7 (BGN/KA/4L) | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AGT angiotensinogen (serpin peptidase inhibitor, clade A, member 8), angiotensin I, angiotensin II, ANHU, SERPINA8 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This product reacts with Angiotensin II and III. Negligible reactivity was observed with Angiotensin I and Angiotensinogen. | |
| Store at 4C in the dark. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu