missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse ANGPTL3 ELISA Kit

Product Code. 18507874
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18507874 96 Tests Pack of 10
15425183 96 Tests 1 set
17175679 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18507874 Supplier Invitrogen™ Supplier No. EMANGPTL3X10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Mouse ANGPTL3 ELISA quantitates Ms ANGPTL3 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms ANGPTL3. Principle of the method The Mouse ANGPLT3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9R182
Assay Range 40.96 to 10,000 pg/mL
Assay Sensitivity 40 pg/mL
Calibration Range 40 pg/mL to 10,000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Proteases, Peptidases & Inhibitors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse 6 Ckine, Activin A, ADAMTS1, Adiponectin, Angiopoietin-3, ANGTL-4 Artemin, CCL28, CD36, Chordin, CRP, E-Cadherin, Epigen, Epiregulin, Fas, Galectin-7, gp130, Granzyme B, Gremlin, IFN-R1, IL-17 B, IL-17 B R, IL-22, MIP-1 b, MMP-2, MMP-3, MMP-10, PDGF-AA, Persephin, sFRP-3, Shh-N, SLAM, TCK-1, TECK, TGFb1, TNF RI, TNF RII, TPO, TRANCE, TremL1, TWEAK, VEGF-B, VEGF-R2, VCAM-1, VEGF.
Sufficient For 96 Assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 30924
Gene Symbol hypl
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Endocrinology
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym RP23-101O16.2, hypl
Target ANGPTL3
Test Time 1 hr. 20 min.
Total Assay Time 4 hr. 45 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.